Assay Details
| Gene Symbol: VPS26C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008274876.2 | XP_008273098.1 | 323 | 77 | ||
| XM_008274877.2 | XP_008273099.1 | 329 | 77 | |||
| XM_017350796.1 | XP_017206285.1 | 327 | 77 | |||
More Information
Set Membership: |
|

VPS26C
Genomic Map