Assay Details
| Gene Symbol: ARL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008270423.2 | XP_008268645.1 | 4 - 5 | 417 | 88 | |
| XM_008270424.2 | XP_008268646.1 | 4 - 5 | 334 | 88 | ||
| XM_017348527.1 | XP_017204016.1 | 4 - 5 | 350 | 88 | ||
More Information
Set Membership: |
|

ARL3
Genomic Map