Assay Details
| Gene Symbol: CES3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711646.3 | XP_002711692.2 | 12 - 13 | 1573 | 91 | |
| XM_017342279.1 | XP_017197768.1 | 12 - 13 | 1552 | 91 | ||
More Information
Set Membership: |
|

CES3
Genomic Map