Assay Details
| Gene Symbol: GPR137B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008268198.2 | XP_008266420.2 | 6 - 7 | 1160 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR137B| Gene Symbol: GPR137B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008268198.2 | XP_008266420.2 | 6 - 7 | 1160 | 63 | |
Set Membership: |
|