Assay Details
| Gene Symbol: LOC100350827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262483.2 | XP_008260705.1 | 11 - 12 | 1463 | 107 | |
| XM_008262484.2 | XP_008260706.1 | 9 - 10 | 1357 | 107 | ||
| XM_017344959.1 | XP_017200448.1 | 12 - 13 | 1567 | 107 | ||
More Information
Set Membership: |
|

LOC100350827
Genomic Map