Assay Details
| Gene Symbol: LOC100350827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262483.2 | XP_008260705.1 | 1 - 2 | 135 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008262483.2 transcripts.
Set Membership: |
|

LOC100350827
Genomic Map