Assay Details
| Gene Symbol: TDGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260522.2 | XP_008258744.1 | 3 - 4 | 689 | 53 | |
| XM_017343777.1 | XP_017199266.1 | 2 - 3 | 287 | 53 | ||
| XM_017343778.1 | XP_017199267.1 | 3 - 4 | 324 | 53 | ||
| XM_017343779.1 | XP_017199268.1 | 3 - 4 | 380 | 53 | ||
More Information
Set Membership: |
|

TDGF1
Genomic Map