Assay Details
| Gene Symbol: HAVCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008255315.2 | XP_008253537.1 | 6 - 7 | 1082 | 73 | |
| XM_008255318.2 | XP_008253540.1 | 5 - 6 | 1005 | 73 | ||
| XM_008255319.2 | XP_008253541.1 | 4 - 5 | 900 | 73 | ||
| XM_017341220.1 | XP_017196709.1 | 6 - 7 | 1080 | 73 | ||
| XM_017341221.1 | XP_017196710.1 | 5 - 6 | 1005 | 73 | ||
| XM_017341222.1 | XP_017196711.1 | 5 - 6 | 980 | 73 | ||
| XM_017341223.1 | XP_017196712.1 | 6 - 7 | 790 | 73 | ||
More Information
Set Membership: |
|

HAVCR1
Genomic Map