Assay Details
| Gene Symbol: NAGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721379.3 | XP_002721425.1 | 877 | 57 | ||
| XM_017350936.1 | XP_017206425.1 | 1095 | 57 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NAGA| Gene Symbol: NAGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721379.3 | XP_002721425.1 | 877 | 57 | ||
| XM_017350936.1 | XP_017206425.1 | 1095 | 57 | |||
Set Membership: |
|