Assay Details
| Gene Symbol: LOC100344938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269427.2 | XP_008267649.1 | 5 - 6 | 1598 | 70 | |
| XM_008269428.2 | XP_008267650.1 | 5 - 6 | 1668 | 70 | ||
| XM_008269429.2 | XP_008267651.1 | 10 - 11 | 2387 | 70 | ||
More Information
Set Membership: |
|

LOC100344938
Genomic Map