Assay Details
| Gene Symbol: IMMP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709031.3 | XP_002709077.1 | 4 - 5 | 363 | 84 | |
| XM_008269310.2 | XP_008267532.1 | 4 - 5 | 443 | 84 | ||
| XM_008269313.2 | XP_008267535.1 | 4 - 5 | 428 | 84 | ||
| XM_008269321.2 | XP_008267543.1 | 4 - 5 | 402 | 84 | ||
| XM_017345714.1 | XP_017201203.1 | 5 - 6 | 684 | 84 | ||
| XM_017345716.1 | XP_017201205.1 | 4 - 5 | 429 | 84 | ||
More Information
Set Membership: |
|

IMMP1L
Genomic Map