Assay Details
| Gene Symbol: KIRREL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264256.2 | XP_008262478.1 | 11 - 12 | 1626 | 58 | |
| XM_008264257.2 | XP_008262479.1 | 11 - 12 | 1578 | 58 | ||
| XM_008264258.2 | XP_008262480.1 | 10 - 11 | 1476 | 58 | ||
More Information
Set Membership: |
|

KIRREL1
Genomic Map