Assay Details
| Gene Symbol: KIRREL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264256.2 | XP_008262478.1 | 14 - 15 | 1958 | 59 | |
| XM_008264257.2 | XP_008262479.1 | 14 - 15 | 1910 | 59 | ||
| XM_008264258.2 | XP_008262480.1 | 13 - 14 | 1808 | 59 | ||
More Information
Set Membership: |
|

KIRREL1
Genomic Map