Assay Details
| Gene Symbol: LOC100358347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008258089.2 | XP_008256311.2 | 1 - 2 | 98 | 54 | |
| XM_017342676.1 | XP_017198165.1 | 1 - 2 | 101 | 54 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008258089.2, XM_017342676.1 transcripts.
Set Membership: |
|

LOC100358347
Genomic Map