Assay Details
| Gene Symbol: LOC100358347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008258089.2 | XP_008256311.2 | 4 - 5 | 1824 | 88 | |
| XM_017342676.1 | XP_017198165.1 | 4 - 5 | 1827 | 88 | ||
More Information
Set Membership: |
|

LOC100358347
Genomic Map