Assay Details
| Gene Symbol: CA5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719890.3 | XP_002719936.1 | 6 - 7 | 788 | 79 | |
| XM_017349800.1 | XP_017205289.1 | 8 - 9 | 1037 | 79 | ||
| XM_017349801.1 | XP_017205290.1 | 8 - 9 | 1000 | 79 | ||
| XM_017349802.1 | XP_017205291.1 | 7 - 8 | 865 | 79 | ||
More Information
Set Membership: |
|

CA5B
Genomic Map