Assay Details
| Gene Symbol: CCN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263390.2 | XP_008261612.1 | 1 - 2 | 298 | 89 | |
| XM_017345323.1 | XP_017200812.1 | 1 - 2 | 132 | 89 | ||
| XM_017345324.1 | XP_017200813.1 | 1 - 2 | 298 | 89 | ||
More Information
Set Membership: |
|

CCN6
Genomic Map