Assay Details
Gene Symbol: LOC103349247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008260875.2 | XP_008259097.1 | 2 - 3 | 564 | 63 | |
XM_008260876.2 | XP_008259098.1 | 2 - 3 | 564 | 63 | ||
XM_017343928.1 | XP_017199417.1 | 2 - 3 | 564 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
