Assay Details
Gene Symbol: LOC103349247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008260875.2 | XP_008259097.1 | 4 - 5 | 796 | 93 | |
XM_008260879.2 | XP_008259101.1 | 3 - 4 | 473 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
