Assay Details
| Gene Symbol: DDX19A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711716.3 | XP_002711762.3 | 3 - 4 | 263 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DDX19A| Gene Symbol: DDX19A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711716.3 | XP_002711762.3 | 3 - 4 | 263 | 86 | |
Set Membership: |
|