Assay Details
| Gene Symbol: SDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272281.2 | XP_008270503.1 | 7 - 8 | 853 | 53 | |
| XM_008272282.2 | XP_008270504.1 | 7 - 8 | 853 | 53 | ||
More Information
Set Membership: |
|

SDS
Genomic Map