Assay Details
| Gene Symbol: DEPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722527.3 | XP_002722573.1 | 197 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002722527.3 transcripts.
Set Membership: |
|

DEPP1
Genomic Map