Assay Details
| Gene Symbol: ALKAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723190.3 | XP_002723236.1 | 461 | 77 | ||
| XM_008251378.2 | XP_008249600.1 | 690 | 77 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALKAL2| Gene Symbol: ALKAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723190.3 | XP_002723236.1 | 461 | 77 | ||
| XM_008251378.2 | XP_008249600.1 | 690 | 77 | |||
Set Membership: |
|