Assay Details
| Gene Symbol: ALKAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008251378.2 | XP_008249600.1 | 766 | 74 | ||
| XM_017339211.1 | XP_017194700.1 | 458 | 74 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALKAL2| Gene Symbol: ALKAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008251378.2 | XP_008249600.1 | 766 | 74 | ||
| XM_017339211.1 | XP_017194700.1 | 458 | 74 | |||
Set Membership: |
|