Assay Details
| Gene Symbol: LOC100353534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262601.2 | XP_008260823.1 | 2 | 2188 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100353534| Gene Symbol: LOC100353534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262601.2 | XP_008260823.1 | 2 | 2188 | 101 | |
Set Membership: |
|