Assay Details
| Gene Symbol: DAXX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262784.2 | XP_008261006.1 | 5 - 6 | 2558 | 58 | |
| XM_017345026.1 | XP_017200515.1 | 6 - 7 | 2187 | 58 | ||
| XM_017345027.1 | XP_017200516.1 | 6 - 7 | 2373 | 58 | ||
| XM_017345028.1 | XP_017200517.1 | 6 - 7 | 2107 | 58 | ||
| XM_017345029.1 | XP_017200518.1 | 6 - 7 | 2184 | 58 | ||
| XM_017345030.1 | XP_017200519.1 | 6 - 7 | 2256 | 58 | ||
| XM_017345031.1 | XP_017200520.1 | 6 - 7 | 2104 | 58 | ||
| XM_017345032.1 | XP_017200521.1 | 6 - 7 | 2138 | 58 | ||
More Information
Set Membership: |
|

DAXX
Genomic Map