Assay Details
| Gene Symbol: MUC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008266760.2 | XP_008264982.1 | 8 - 9 | 1297 | 73 | |
| XM_008266761.2 | XP_008264983.1 | 8 - 9 | 1297 | 73 | ||
More Information
Set Membership: |
|

MUC13
Genomic Map