Assay Details
| Gene Symbol: SAPCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714311.3 | XP_002714357.1 | 2 - 3 | 480 | 63 | |
| XM_008262592.2 | XP_008260814.1 | 2 - 3 | 480 | 63 | ||
More Information
Set Membership: |
|

SAPCD1
Genomic Map