Assay Details
| Gene Symbol: LOC100351062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008274285.2 | XP_008272507.1 | 2 - 3 | 698 | 70 | |
| XM_017350399.1 | XP_017205888.1 | 2 - 3 | 698 | 70 | ||
More Information
Set Membership: |
|

LOC100351062
Genomic Map