Assay Details
| Gene Symbol: LOC100351062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008274285.2 | XP_008272507.1 | 3 - 4 | 783 | 102 | |
| XM_017350399.1 | XP_017205888.1 | 3 - 4 | 783 | 102 | ||
| XM_017350401.1 | XP_017205890.1 | 2 - 3 | 693 | 102 | ||
| XM_017350406.1 | XP_017205895.1 | 2 - 3 | 693 | 102 | ||
More Information
Set Membership: |
|

LOC100351062
Genomic Map