Assay Details
| Gene Symbol: LOC100351062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008274285.2 | XP_008272507.1 | 6 - 7 | 1032 | 73 | |
| XM_017350399.1 | XP_017205888.1 | 5 - 6 | 1008 | 73 | ||
| XM_017350401.1 | XP_017205890.1 | 5 - 6 | 942 | 73 | ||
| XM_017350406.1 | XP_017205895.1 | 4 - 5 | 918 | 73 | ||
More Information
Set Membership: |
|

LOC100351062
Genomic Map