Assay Details
| Gene Symbol: MOGAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712499.3 | XP_002712545.1 | 2 - 3 | 532 | 110 | |
| XM_017343157.1 | XP_017198646.1 | 2 - 3 | 541 | 110 | ||
More Information
Set Membership: |
|

MOGAT1
Genomic Map