Assay Details
| Gene Symbol: SKIL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017346914.1 | XP_017202403.1 | 6 - 7 | 2577 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SKIL| Gene Symbol: SKIL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017346914.1 | XP_017202403.1 | 6 - 7 | 2577 | 65 | |
Set Membership: |
|