Assay Details
| Gene Symbol: PIPOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718915.3 | XP_002718961.1 | 2 - 3 | 347 | 67 | |
| XM_017348942.1 | XP_017204431.1 | 2 - 3 | 351 | 67 | ||
More Information
Set Membership: |
|

PIPOX
Genomic Map