Assay Details
| Gene Symbol: CHST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709072.3 | XP_002709118.1 | 1 - 2 | 184 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002709072.3 transcripts.
Set Membership: |
|

CHST1
Genomic Map