Assay Details
| Gene Symbol: LOC100343171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721365.3 | XP_002721411.1 | 136 | 61 | ||
| GenBank mRNA | JQ173124.1 | - | 122 | 61 | ||
| JQ173125.1 | - | 122 | 61 | |||
| JQ173126.1 | - | 122 | 61 | |||
| JQ173127.1 | - | 122 | 61 | |||
More Information
Set Membership: |
|

LOC100343171
Genomic Map