Assay Details
| Gene Symbol: TSGA10IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017348799.1 | XP_017204288.1 | 196 | 51 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSGA10IP| Gene Symbol: TSGA10IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017348799.1 | XP_017204288.1 | 196 | 51 | ||
Set Membership: |
|