Assay Details
| Gene Symbol: PTER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008268303.2 | XP_008266525.1 | 4 - 5 | 997 | 73 | |
| XM_008268304.2 | XP_008266526.1 | 4 - 5 | 976 | 73 | ||
More Information
Set Membership: |
|

PTER
Genomic Map