Assay Details
| Gene Symbol: ANGPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008255827.2 | XP_008254049.1 | 2 - 3 | 1128 | 70 | |
| XM_008255828.2 | XP_008254050.1 | 2 - 3 | 1125 | 70 | ||
More Information
Set Membership: |
|

ANGPT1
Genomic Map