Assay Details
| Gene Symbol: METTL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718874.3 | XP_002718920.1 | 9 - 10 | 1207 | 68 | |
| XM_008270913.2 | XP_008269135.1 | 10 - 11 | 1370 | 68 | ||
| XM_008270914.2 | XP_008269136.1 | 9 - 10 | 1281 | 68 | ||
| XM_017348900.1 | XP_017204389.1 | 7 - 8 | 808 | 68 | ||
| XM_017348901.1 | XP_017204390.1 | 8 - 9 | 1078 | 68 | ||
More Information
Set Membership: |
|

METTL16
Genomic Map