Assay Details
| Gene Symbol: LOC100009451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008252470.1 | XP_008250692.1 | 680 | 79 | ||
| GenBank mRNA | AB016251.1 | - | 219 | 79 | ||
More Information
Set Membership: |
|

LOC100009451
Genomic Map