Assay Details
| Gene Symbol: C1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712749.3 | XP_002712795.1 | 11 | 1954 | 105 | |
| XM_008259770.2 | XP_008257992.1 | 10 | 1982 | 105 | ||
| GenBank mRNA | AF108768.1 | - | 1 | 119 | 105 | |
More Information
Set Membership: |
|

C1R
Genomic Map