Assay Details
| Gene Symbol: COX4I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361489.1 | NP_001348418.1 | 443 | 75 | ||
| NM_001361490.1 | NP_001348419.1 | 720 | 75 | |||
| GenBank mRNA | AF198088.1 | - | 420 | 75 | ||
| EB373756.1 | - | 428 | 75 | |||
More Information
Set Membership: |
|

COX4I1
Genomic Map