Assay Details
| Gene Symbol: RXRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714483.3 | XP_002714529.1 | 9 | 1387 | 57 | |
| XM_008262773.2 | XP_008260995.1 | 9 | 1399 | 57 | ||
| XM_008262774.2 | XP_008260996.1 | 9 | 1336 | 57 | ||
| GenBank mRNA | DQ102475.1 | - | 1 | 38 | 57 | |
More Information
Set Membership: |
|

RXRB
Genomic Map