Assay Details
| Gene Symbol: IDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712618.3 | XP_002712664.1 | 7 - 8 | 1087 | 74 | |
| XM_017343066.1 | XP_017198555.1 | 5 - 6 | 851 | 74 | ||
| GenBank mRNA | DQ403080.1 | - | 5 - 6 | 767 | 74 | |
More Information
Set Membership: |
|

IDH1
Genomic Map