Assay Details
| Gene Symbol: ATP5F1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711084.3 | XP_002711130.3 | 4 - 5 | 648 | 101 | |
| GenBank mRNA | DQ403105.1 | - | 3 - 4 | 358 | 101 | |
More Information
Set Membership: |
|

ATP5F1B
Genomic Map