Assay Details
| Gene Symbol: CS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711075.3 | XP_002711121.1 | 8 - 9 | 1091 | 70 | |
| GenBank mRNA | DQ403130.1 | - | 2 - 3 | 252 | 70 | |
More Information
Set Membership: |
|

CS
Genomic Map