Assay Details
| Gene Symbol: MMP17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002724095.2 | XP_002724141.2 | 1118 | 64 | ||
| GenBank mRNA | EF472905.1 | - | 281 | 64 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MMP17| Gene Symbol: MMP17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002724095.2 | XP_002724141.2 | 1118 | 64 | ||
| GenBank mRNA | EF472905.1 | - | 281 | 64 | ||
Set Membership: |
|