Assay Details
| Gene Symbol: TLR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260758.2 | XP_008258980.1 | 7 - 8 | 943 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TLR9| Gene Symbol: TLR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260758.2 | XP_008258980.1 | 7 - 8 | 943 | 75 | |
Set Membership: |
|