Assay Details
| Gene Symbol: MYLK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721559.3 | XP_002721605.1 | 2002 | 57 | ||
| XM_008248378.2 | XP_008246600.1 | 1558 | 57 | |||
| XM_008248379.2 | XP_008246601.1 | 1423 | 57 | |||
| XM_008248380.2 | XP_008246602.1 | 1503 | 57 | |||
More Information
Set Membership: |
|

MYLK3
Genomic Map